The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_026796.1 | 3 - 4 | 223 | 63 | |
NR_026797.1 | 2 - 3 | 161 | 63 | ||
GenBank mRNA | BU171486.1 | - | 3 - 4 | 223 | 63 |
BU178088.1 | - | 2 - 3 | 173 | 63 |