The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_026850.1 | 2 - 3 | 1377 | 90 | |
GenBank mRNA | AK096109.1 | - | 1 - 2 | 929 | 90 |
AL833686.1 | - | 2 - 3 | 1287 | 90 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | eukaryotic translation initiation factor 3 subunit I | EIF3I | EIF3S2, PRO2242, TRIP-1, TRIP1, eIF3-beta, eIF3-p36 | -7.7kb | Anti-sense | Chr. - 32221584 - 32231604 on Build GRCh38 |
Downstream | family with sequence similarity 167 member B | FAM167B | C1orf90 | 6.6kb | Anti-sense | Chr. - 32247217 - 32248860 on Build GRCh38 |