The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_026850.1 | 4 - 5 | 1757 | 101 | |
GenBank mRNA | AK096109.1 | - | 3 - 4 | 1309 | 101 |
AL833686.1 | - | 4 - 5 | 1570 | 101 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | eukaryotic translation initiation factor 3 subunit I | EIF3I | EIF3S2, PRO2242, TRIP-1, TRIP1, eIF3-beta, eIF3-p36 | -3.1kb | Anti-sense | Chr. - 32221584 - 32231604 on Build GRCh38 |
Downstream | family with sequence similarity 167 member B | FAM167B | C1orf90 | 12.0kb | Anti-sense | Chr. - 32247217 - 32248860 on Build GRCh38 |