The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_026893.1 | 5 - 6 | 602 | 65 | |
XM_017019507.1 | XP_016874996.1 | 5 - 6 | 1783 | 65 | |
XM_017019508.1 | XP_016874997.1 | 6 - 7 | 1694 | 65 | |
XM_017019509.1 | XP_016874998.1 | 4 - 5 | 703 | 65 | |
XM_017019510.1 | XP_016874999.1 | 5 - 6 | 1132 | 65 | |
XM_017019511.1 | XP_016875000.1 | 5 - 6 | 1301 | 65 | |
XM_017019512.1 | XP_016875001.1 | 5 - 6 | 657 | 65 | |
XM_017019514.1 | XP_016875003.1 | 5 - 6 | 1783 | 65 | |
XM_017019515.1 | XP_016875004.1 | 5 - 6 | 1129 | 65 | |
XM_017019517.1 | XP_016875006.1 | 5 - 6 | 1783 | 65 | |
XM_017019518.1 | XP_016875007.1 | 5 - 6 | 1783 | 65 | |
XM_017019519.1 | XP_016875008.1 | 5 - 6 | 1783 | 65 | |
XM_017019522.1 | XP_016875011.1 | 4 - 5 | 507 | 65 | |
XM_017019526.1 | XP_016875015.1 | 4 - 5 | 507 | 65 | |
XM_017019527.1 | XP_016875016.1 | 4 - 5 | 507 | 65 | |
XM_017019529.1 | XP_016875018.1 | 4 - 5 | 918 | 65 | |
XM_017019530.1 | XP_016875019.1 | 3 - 4 | 823 | 65 | |
GenBank mRNA | AK095191.1 | - | 5 - 6 | 602 | 65 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | transcription factor CP2 | TFCP2 | LBP1C, LSF, LSF1D, SEF, TFCP2C | -24.6kb | Sense | Chr. - 51093756 - 51173143 on Build GRCh38 |
Downstream | DAZ associated protein 2 | DAZAP2 | PRTB | 40.0kb | Anti-sense | Chr. - 51238724 - 51246717 on Build GRCh38 |