Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HLA-F-AS1
Entrez Gene ID
285830
Gene Name
HLA-F antisense RNA 1
Gene Aliases
-
Chromosome Location
Chr.6: 29726601 - 29749049 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_026972.1 5 - 6 1373 94
GenBank mRNA AK092748.1 - 5 - 6 1373 94

View Full Product Details