Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YBX3P1
Entrez Gene ID
440359
Gene Name
Y-box binding protein 3 pseudogene 1
Gene Aliases
CSDAP1
Chromosome Location
Chr.16: 31567767 - 31569524 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_027011.1 1 - 1 1649 112
GenBank mRNA AI183563.1 - 1 - 1 112


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CD243796.1 - 1 139 Chr. 16 - 31563931 - 31568011 [-] on Build GRCh38 111
GenBankmRNA HY223748.1 - 1 Chr. 16 - 31567802 - 31568263 [+] on Build GRCh38 112


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream FSHD region gene 2 family member K, pseudogene FRG2KP Sense Chr. - 31563222 - 31567715 on Build GRCh38
Downstream clustered mitochondria homolog pseudogene 3 CLUHP3 C16orf67, KIAA0664L3, KIAA0664P3, URLC3 131.1kb Anti-sense Chr. - 31700613 - 31707437 on Build GRCh38

View Full Product Details