Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LINC00628
- Entrez Gene ID
-
127841
- Gene Name
- long intergenic non-protein coding RNA 628
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 204368430 - 204369719 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027022.1 |
|
1 - 1 |
854 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ribosomal protein L21 pseudogene 19 |
RPL21P19 |
RPL21_7_151 |
-21.1kb
|
Sense |
Chr. - 204346757 - 204347298 on Build GRCh38 |
Downstream |
protein phosphatase 1 regulatory subunit 15B |
PPP1R15B |
CREP, MSSGM2 |
30.6kb
|
Sense |
Chr. - 204398684 - 204411880 on Build GRCh38 |
View Full Product Details