Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- GUSBP1
- Entrez Gene ID
-
728411
- Gene Name
- glucuronidase, beta pseudogene 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 21459480 - 21589372 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027026.1 |
|
4 - 5 |
386 |
83 |
GenBank mRNA |
AK095656.1 |
- |
4 - 5 |
310 |
83 |
DA358891.1 |
- |
4 - 5 |
386 |
83 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA112079.1 |
- |
4 - 5 |
385 |
|
Chr. 5 - 21459483 - 21585672 [+] on Build GRCh38 |
83 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105374676 |
LOC105374676 |
|
-564.0kb
|
Sense |
Chr. - 20611833 - 20937711 on Build GRCh38 |
Downstream |
cadherin 12 |
CDH12 |
CDHB |
161.5kb
|
Anti-sense |
Chr. - 21750191 - 22853622 on Build GRCh38 |
View Full Product Details