Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RPL21P44
- Entrez Gene ID
-
402176
- Gene Name
- ribosomal protein L21 pseudogene 44
- Gene Aliases
-
RPL21_17_477
- Chromosome Location
-
Chr.4: 53985486 - 53987282 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027153.1 |
|
1 - 1 |
854 |
74 |
GenBank mRNA |
BC057822.1 |
- |
2 - 2 |
825 |
74 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC100506444 |
LOC100506444 |
|
-251.1kb
|
Anti-sense |
Chr. - 53695901 - 53734358 on Build GRCh38 |
Downstream |
cysteine rich hydrophobic domain 2 |
CHIC2 |
BTL |
21.1kb
|
Sense |
Chr. - 53996335 - 54064648 on Build GRCh38 |
View Full Product Details