Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TATDN1
Entrez Gene ID
83940
Gene Name
TatD DNase domain containing 1
Gene Aliases
CDA11
Chromosome Location
Chr.8: 124488494 - 124539088 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146160.1 NP_001139632.1 4 - 5 305 119
NM_001317889.1 NP_001304818.1 6 - 7 421 119
NM_001317890.1 NP_001304819.1 4 - 5 404 119
NM_032026.3 NP_114415.1 6 - 7 421 119
NR_027427.1 6 - 7 421 119
XM_006716666.3 XP_006716729.1 6 - 7 441 119
XM_006716667.3 XP_006716730.1 6 - 7 486 119
XM_006716668.3 XP_006716731.1 6 - 7 499 119
XM_006716669.3 XP_006716732.1 4 - 5 383 119
XM_006716670.3 XP_006716733.1 5 - 6 342 119
XM_006716671.3 XP_006716734.1 5 - 6 420 119
XM_011517331.2 XP_011515633.1 5 - 6 337 119
XM_017013895.1 XP_016869384.1 6 - 7 431 119
XM_017013896.1 XP_016869385.1 6 - 7 486 119
XM_017013897.1 XP_016869386.1 5 - 6 407 119
XM_017013898.1 XP_016869387.1 5 - 6 344 119
XM_017013899.1 XP_016869388.1 6 - 7 417 119
GenBank mRNA AF212250.1 - 6 - 7 404 119
AK312204.1 - 6 - 7 417 119
AY071865.1 - 4 - 5 294 119
BC064964.1 - 6 - 7 405 119
BG532054.1 - 4 - 5 394 119
BX332564.2 - 6 - 7 403 119

View Full Product Details