Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SNAR-D
- Entrez Gene ID
-
100170227
- Gene Name
- small ILF3/NF90-associated RNA D
- Gene Aliases
-
-
- Chromosome Location
-
Chr.19: 50140202 - 50140320 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_024243.1 |
|
1 - 1 |
56 |
78 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
small ILF3/NF90-associated RNA B2 |
SNAR-B2 |
|
-965.0
|
Sense |
Chr. - 50139118 - 50139237 on Build GRCh38 |
Downstream |
IZUMO family member 2 |
IZUMO2 |
C19orf41, PLAL6978, PRO21961, SCRL |
12.2kb
|
Sense |
Chr. - 50152548 - 50163281 on Build GRCh38 |
View Full Product Details