The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323638.1 | NP_001310567.1 | 2 - 3 | 310 | 61 |
NM_001323642.1 | NP_001310571.1 | 1 - 2 | 354 | 61 | |
NM_001323646.1 | NP_001310575.1 | 1 - 2 | 354 | 61 | |
GenBank mRNA | AK309331.1 | - | 1 - 2 | 354 | 61 |
AK311372.1 | - | 1 - 2 | 300 | 61 | |
BX647794.1 | - | 2 - 3 | 311 | 61 |