Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LOC441666
- Entrez Gene ID
-
441666
- Gene Name
- zinc finger protein 91 pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 42331866 - 42368045 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_024380.1 |
|
1 - 2 |
229 |
103 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA322429.1 |
- |
1 - 2 |
158 |
|
Chr. 10 - 42337880 - 42367974 [-] on Build GRCh38 |
103 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ACTR3B pseudogene 5 |
ACTR3BP5 |
FKSG74 |
-3.6mb
|
Anti-sense |
Chr. - 38696596 - 38698241 on Build GRCh38 |
Downstream |
cyclin Y-like 2 (pseudogene) |
CCNYL2 |
C10orf21, CCNYL2P, bA178A10.2 |
40.1kb
|
Sense |
Chr. - 42408174 - 42472240 on Build GRCh38 |
View Full Product Details