Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HINT1
Entrez Gene ID
3094
Gene Name
histidine triad nucleotide binding protein 1
Gene Aliases
HINT, NMAN, PKCI-1, PRKCNH1
Chromosome Location
Chr.5: 131159283 - 131165348 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_024611.2 1 - 2 291 95
NR_073488.1 1 - 2 291 95
GenBank mRNA CD639104.1 - 1 - 2 166 95
CK002748.1 - 1 - 2 279 95


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BG504755.1 - 1 - 2 164 Chr. 5 - 131159285 - 131165220 [-] on Build GRCh38 95
GenBankmRNA DA235754.1 - 1 - 2 199 Chr. 5 - 131162291 - 131165256 [-] on Build GRCh38 95
GenBankmRNA HY182087.1 - 1 - 2 175 Chr. 5 - 131159413 - 131165232 [-] on Build GRCh38 95
GenBankmRNA BP231584.1 - 1 - 2 197 Chr. 5 - 131159329 - 131165254 [-] on Build GRCh38 95
GenBankmRNA HY376070.1 - 2 - 3 Chr. 5 - 131159299 - 131165118 [+] on Build GRCh38 95
GenBankmRNA BE874740.1 - 1 - 2 182 Chr. 5 - 131159292 - 131165239 [-] on Build GRCh38 95
GenBankmRNA EL949508.1 - 1 - 2 143 Chr. 5 - 131159336 - 131165198 [-] on Build GRCh38 95
GenBankmRNA BU533696.1 - 1 - 2 201 Chr. 5 - 131159285 - 131165255 [-] on Build GRCh38 95


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream chondroitin sulfate synthase 3 CHSY3 CHSY2, CSS3 -975.9kb Anti-sense Chr. - 129903415 - 130186637 on Build GRCh38
Downstream LYR motif containing 7 LYRM7 C5orf31, MC3DN8, MZM1L 5.6kb Anti-sense Chr. - 131170914 - 131205426 on Build GRCh38

View Full Product Details