Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FAM99A
- Entrez Gene ID
-
387742
- Gene Name
- family with sequence similarity 99 member A (non-protein coding)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 1665599 - 1667856 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026643.1 |
|
3 - 3 |
1255 |
78 |
GenBank mRNA |
AK124823.1 |
- |
1 - 1 |
1859 |
78 |
CR627417.1 |
- |
3 - 3 |
1282 |
78 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB306826.1 |
- |
1 |
|
|
Chr. 11 - 1667321 - 1667855 [-] on Build GRCh38 |
78 |
GenBankmRNA |
DB321251.1 |
- |
1 |
|
|
Chr. 11 - 1667315 - 1667865 [-] on Build GRCh38 |
78 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
keratin associated protein 5-5 |
KRTAP5-5 |
KRTAP5-11, KRTAP5.5 |
-35.8kb
|
Sense |
Chr. - 1629803 - 1630930 on Build GRCh38 |
Downstream |
family with sequence similarity 99 member B (non-protein coding) |
FAM99B |
|
15.4kb
|
Anti-sense |
Chr. - 1683270 - 1685629 on Build GRCh38 |
View Full Product Details