The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_212469.1 | NP_997634.1 | 2 - 3 | 675 | 64 |
GenBank mRNA | AI809679.1 | - | 3 - 4 | 329 | 64 |