Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PLGLB1
- Entrez Gene ID
-
5343
- Gene Name
- plasminogen-like B1
- Gene Aliases
-
PLGL, PLGP1, PRGB, PRP-B
- Chromosome Location
-
Chr.2: 87010464 - 87021846 on Build GRCh38
- Gene Symbol
- PLG
- Entrez Gene ID
-
5340
- Gene Name
- plasminogen
- Gene Aliases
-
-
- Chromosome Location
-
Chr.6: 160702193 - 160754054 on Build GRCh38
- Gene Symbol
- PLGLA
- Entrez Gene ID
-
285189
- Gene Name
- plasminogen-like A (pseudogene)
- Gene Aliases
-
PLGLA1, PLGP2, PRGA
- Chromosome Location
-
Chr.2: 106382114 - 106391395 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003506.3 |
|
1 - 2 |
106 |
126 |
GenBank mRNA |
DA634686.1 |
- |
1 - 2 |
106 |
127 |
- Gene Symbol
- PLGLB2
- Entrez Gene ID
-
5342
- Gene Name
- plasminogen-like B2
- Gene Aliases
-
PLGP1
- Chromosome Location
-
Chr.2: 87748087 - 87759476 on Build GRCh38
View Full Product Details