The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016334.3 | NP_057418.1 | 10 - 11 | 1053 | 84 |
XM_005277401.3 | XP_005277458.1 | 10 - 11 | 1026 | 84 | |
XM_005277402.4 | XP_005277459.1 | 10 - 11 | 1190 | 84 | |
XM_005277404.3 | XP_005277461.1 | 10 - 11 | 1046 | 84 | |
XM_006711377.3 | XP_006711440.1 | 11 - 12 | 1089 | 84 | |
XM_006711378.2 | XP_006711441.1 | 11 - 12 | 1092 | 84 | |
XM_011509610.2 | XP_011507912.1 | 11 - 12 | 1086 | 84 | |
XM_011509612.2 | XP_011507914.1 | 11 - 12 | 1047 | 84 | |
XM_017001448.1 | XP_016856937.1 | 11 - 12 | 1589 | 84 | |
GenBank mRNA | AK299995.1 | - | 10 - 11 | 1024 | 84 |
AL133020.1 | - | 10 - 11 | 1554 | 84 | |
CD102528.1 | - | 5 - 6 | 418 | 84 | |
KJ534849.1 | - | 10 - 11 | 915 | 84 | |
KJ534850.1 | - | 10 - 11 | 909 | 84 | |
KJ534851.1 | - | 10 - 11 | 840 | 84 | |
KJ535070.1 | - | 11 - 12 | 1237 | 84 | |
U78723.1 | - | 10 - 11 | 1012 | 84 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001097612.1 | NP_001091081.1 | 10 - 11 | 1083 | 84 |
NM_001097613.2 | NP_001091082.2 | 11 - 12 | 1129 | 84 | |
NR_036541.1 | 10 - 11 | 1103 | 84 | ||
XM_006711492.3 | XP_006711555.1 | 10 - 11 | 1053 | 84 | |
XM_006711493.3 | XP_006711556.1 | 11 - 12 | 1061 | 84 | |
XM_011509908.2 | XP_011508210.1 | 11 - 12 | 1099 | 84 | |
XM_011509909.2 | XP_011508211.1 | 10 - 11 | 1311 | 84 | |
GenBank mRNA | AB097024.1 | - | 10 - 11 | 1024 | 84 |
AB097025.1 | - | 10 - 11 | 1024 | 84 | |
AF132947.1 | - | 10 - 11 | 1008 | 84 | |
AK001354.1 | - | 3 - 4 | 1821 | 84 | |
AK021758.1 | - | 10 - 11 | 1073 | 84 | |
AK094228.1 | - | 1 - 2 | 1024 | 84 | |
AK301939.1 | - | 10 - 11 | 930 | 84 | |
AK316371.1 | - | 11 - 12 | 1041 | 84 | |
AY358966.1 | - | 10 - 11 | 1058 | 84 | |
BC003187.1 | - | 10 - 11 | 986 | 84 | |
BC067816.1 | - | 10 - 11 | 1042 | 84 | |
BC146880.1 | - | 10 - 11 | 1004 | 84 | |
BT006679.1 | - | 10 - 11 | 909 | 84 | |
KJ534847.1 | - | 10 - 11 | 909 | 84 | |
KJ534848.1 | - | 10 - 11 | 915 | 84 | |
KJ535029.1 | - | 9 - 10 | 909 | 84 | |
KJ535049.1 | - | 10 - 11 | 929 | 84 |