Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANO6
Entrez Gene ID
196527
Gene Name
anoctamin 6
Gene Aliases
BDPLT7, SCTS, TMEM16F
Chromosome Location
Chr.12: 45215987 - 45440404 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025356.2 NP_001020527.2 17 - 18 2550 104
NM_001142678.1 NP_001136150.1 17 - 18 2397 104
NM_001142679.1 NP_001136151.1 17 - 18 2550 104
NM_001204803.1 NP_001191732.1 18 - 19 2613 104
XM_005268706.4 XP_005268763.1 17 - 18 2504 104
XM_005268707.3 XP_005268764.1 16 - 17 2424 104
GenBank mRNA AK126409.1 - 17 - 18 2631 104
AK302819.1 - 18 - 19 2407 104
AK303996.1 - 17 - 18 2481 104
AK310641.1 - 17 - 18 2517 104
AL832340.1 - 17 - 18 2397 104
BC098410.1 - 17 - 18 2417 104
BC136445.1 - 18 - 19 2306 104

View Full Product Details