Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMCO2
Entrez Gene ID
341346
Gene Name
single-pass membrane protein with coiled-coil domains 2
Gene Aliases
C12orf70
Chromosome Location
Chr.12: 27446482 - 27506911 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145010.1 NP_001138482.1 2 - 3 313 71
XM_011520634.2 XP_011518936.1 2 - 3 491 71
XM_011520636.2 XP_011518938.1 2 - 3 491 71

View Full Product Details