The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145010.1 | NP_001138482.1 | 3 - 4 | 411 | 73 |
XM_011520634.2 | XP_011518936.1 | 3 - 4 | 589 | 73 | |
XM_011520636.2 | XP_011518938.1 | 3 - 4 | 589 | 73 |