The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145010.1 | NP_001138482.1 | 5 - 6 | 679 | 99 |
XM_011520634.2 | XP_011518936.1 | 5 - 6 | 857 | 99 | |
XM_011520635.2 | XP_011518937.1 | 3 - 4 | 356 | 99 | |
XM_017019254.1 | XP_016874743.1 | 3 - 4 | 613 | 99 |