Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HTR3D
Entrez Gene ID
200909
Gene Name
5-hydroxytryptamine receptor 3D
Gene Aliases
5HT3D
Chromosome Location
Chr.3: 184031544 - 184039369 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145143.1 NP_001138615.1 4 - 5 366 66

View Full Product Details