Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REG4
Entrez Gene ID
83998
Gene Name
regenerating family member 4
Gene Aliases
GISP, REG-IV, RELP
Chromosome Location
Chr.1: 119794018 - 119811580 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159352.1 NP_001152824.1 3 - 4 504 84
NM_001159353.1 NP_001152825.1 2 - 3 330 84
NM_032044.3 NP_114433.1 2 - 3 330 84
GenBank mRNA AF254415.1 - 2 - 3 245 84
AF345934.1 - 2 - 3 198 84
AY007243.1 - 2 - 3 186 84
AY126670.1 - 3 - 4 504 84
AY126671.1 - 2 - 3 330 84
AY126672.1 - 2 - 3 330 84
BC017089.2 - 2 - 3 202 84

View Full Product Details