Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI174727.1 - 1 46 Chr. 13 - 41359115 - 41359719 [-] on Build GRCh38 99
GenBankmRNA AF119881.1 - 1 46 Chr. 13 - 41358348 - 41359719 [-] on Build GRCh38 99


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream N(alpha)-acetyltransferase 16, NatA auxiliary subunit NAA16 NARG1L Anti-sense Chr. - 41311205 - 41377031 on Build GRCh38
Downstream olfactory receptor family 7 subfamily E member 37 pseudogene OR7E37P OR7E48P, OST193, hg533 82.8kb Anti-sense Chr. - 41442564 - 41443766 on Build GRCh38

View Full Product Details