Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG195702.1 |
- |
1 |
635 |
|
Chr. 4 - 151171229 - 151171934 [-] on Build GRCh38 |
88 |
GenBankmRNA |
AK000802.1 |
- |
1 |
2052 |
|
Chr. 4 - 151171206 - 151173369 [-] on Build GRCh38 |
88 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
SH3 domain containing 19 |
SH3D19 |
EBP, EVE1, Kryn, SH3P19 |
|
Sense |
Chr. - 151120281 - 151228030 on Build GRCh38 |
Downstream |
protease, serine 48 |
PRSS48 |
ESSPL |
103.4kb
|
Anti-sense |
Chr. - 151276482 - 151291856 on Build GRCh38 |
View Full Product Details