Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AK021574.1 |
- |
1 |
1250 |
|
Chr. 3 - 72927269 - 72928800 [+] on Build GRCh38 |
74 |
GenBankmRNA |
AU143791.1 |
- |
1 |
|
|
Chr. 3 - 72928245 - 72928785 [-] on Build GRCh38 |
74 |
GenBankmRNA |
AU144536.1 |
- |
1 |
|
|
Chr. 3 - 72928240 - 72928800 [-] on Build GRCh38 |
74 |
GenBankmRNA |
AA493671.1 |
- |
1 |
|
|
Chr. 3 - 72928409 - 72928613 [-] on Build GRCh38 |
74 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
glucoside xylosyltransferase 2 |
GXYLT2 |
GLT8D4 |
|
Sense |
Chr. - 72848522 - 72975374 on Build GRCh38 |
Downstream |
protein phosphatase 4 regulatory subunit 2 |
PPP4R2 |
PP4R2 |
68.2kb
|
Sense |
Chr. - 72996743 - 73069201 on Build GRCh38 |
View Full Product Details