Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA M90754.1 - 1 1349 Chr. X - 88496374 - 88498163 [+] on Build GRCh38 109


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream ribosomal protein SA pseudogene 15 RPSAP15 LAMR1P15, LAMRL5, RPSA_25_1791 -792.1kb Anti-sense Chr. - 87703273 - 87704306 on Build GRCh38
Downstream CPX chromosome region, candidate 1 CPXCR1 CT77 249.1kb Sense Chr. - 88747225 - 88754785 on Build GRCh38

View Full Product Details