Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AF085888.1 - 1 453 Chr. 9 - 88450506 - 88451122 [+] on Build GRCh38 65
GenBankmRNA AA252232.1 - 1 Chr. 9 - 88450670 - 88451113 [-] on Build GRCh38 65
GenBankmRNA H30735.1 - 1 Chr. 9 - 88450766 - 88451095 [-] on Build GRCh38 65


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream SPATA31 subfamily C member 2 SPATA31C2 FAM75C2 -315.5kb Anti-sense Chr. - 88129305 - 88134985 on Build GRCh38
Downstream nucleoredoxin-like 2 NXNL2 C9orf121, RDCVF2 84.0kb Sense Chr. - 88535088 - 88584510 on Build GRCh38

View Full Product Details