Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
R05809.1 |
- |
|
|
|
Chr. - - [-] on Build |
92 |
GenBankmRNA |
AF085924.1 |
- |
1 |
454 |
|
Chr. 3 - 73048473 - 73049094 [+] on Build GRCh38 |
92 |
GenBankmRNA |
H62565.1 |
- |
1 |
|
|
Chr. 3 - 73048788 - 73049086 [-] on Build GRCh38 |
92 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
glucoside xylosyltransferase 2 |
GXYLT2 |
GLT8D4 |
-73.1kb
|
Sense |
Chr. - 72848522 - 72975374 on Build GRCh38 |
Downstream |
endogenous Bornavirus-like nucleoprotein 2 |
EBLN2 |
EBLN-2 |
12.6kb
|
Sense |
Chr. - 73061659 - 73063337 on Build GRCh38 |
View Full Product Details