Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SSH2
- Entrez Gene ID
-
85464
- Gene Name
- slingshot protein phosphatase 2
- Gene Aliases
-
SSH-2, SSH-2L
- Chromosome Location
-
Chr.17: 29625938 - 29930239 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF086010.1 |
- |
1 - 1 |
450 |
74 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
H97560.1 |
- |
1 |
|
|
Chr. 17 - 29644799 - 29645213 [+] on Build GRCh38 |
74 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
coronin 6 |
CORO6 |
|
-21.9kb
|
Sense |
Chr. - 29614756 - 29622912 on Build GRCh38 |
Downstream |
EF-hand calcium binding domain 5 |
EFCAB5 |
|
284.5kb
|
Anti-sense |
Chr. - 29929856 - 30108452 on Build GRCh38 |
View Full Product Details