Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
W73662.1 |
- |
|
214 |
|
Chr. - - [-] on Build |
69 |
GenBankmRNA |
AF086279.1 |
- |
1 |
250 |
|
Chr. 14 - 55915727 - 55916326 [+] on Build GRCh38 |
69 |
GenBankmRNA |
BX101908.1 |
- |
1 |
247 |
|
Chr. 14 - 55915730 - 55916306 [+] on Build GRCh38 |
69 |
GenBankmRNA |
W69547.1 |
- |
|
214 |
|
Chr. - - [-] on Build |
69 |
GenBankmRNA |
W69463.1 |
- |
|
|
|
Chr. - - [-] on Build |
69 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 520 |
LINC00520 |
C14orf34 |
-119.1kb
|
Anti-sense |
Chr. - 55781135 - 55796674 on Build GRCh38 |
Downstream |
pellino E3 ubiquitin protein ligase family member 2 |
PELI2 |
|
202.0kb
|
Sense |
Chr. - 56118330 - 56301586 on Build GRCh38 |
View Full Product Details