Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AF087971.1 - 1 347 Chr. 3 - 52625742 - 52626434 [-] on Build GRCh38 74
GenBankmRNA H81077.1 - 1 Chr. 3 - 52625757 - 52626154 [+] on Build GRCh38 74
GenBankmRNA BX097388.1 - 1 346 Chr. 3 - 52625742 - 52626433 [-] on Build GRCh38 74


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream small integral membrane protein 4 SMIM4 C3orf78 -85.2kb Anti-sense Chr. - 52535986 - 52540570 on Build GRCh38
Downstream G protein nucleolar 3 GNL3 C77032, E2IG3, NNP47, NS 59.5kb Anti-sense Chr. - 52685920 - 52694497 on Build GRCh38

View Full Product Details