Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AF088003.1 - 1 152 Chr. 12 - 50157424 - 50158070 [+] on Build GRCh38 78
GenBankmRNA N48540.1 - 1 150 Chr. 12 - 50157427 - 50157859 [+] on Build GRCh38 78
GenBankmRNA BX096676.1 - 1 152 Chr. 12 - 50157424 - 50158075 [+] on Build GRCh38 78


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream ceramide synthase 5 CERS5 LASS5, Trh4 Anti-sense Chr. - 50129306 - 50167533 on Build GRCh38
Downstream LIM domain and actin binding 1 LIMA1 EPLIN, SREBP3 17.7kb Anti-sense Chr. - 50175780 - 50283570 on Build GRCh38

View Full Product Details