Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- METTL15
- Entrez Gene ID
-
196074
- Gene Name
- methyltransferase like 15
- Gene Aliases
-
METT5D1
- Chromosome Location
-
Chr.11: 28108251 - 28527041 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AJ131606.1 |
- |
1 - 1 |
152 |
81 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
kinesin family member 18A |
KIF18A |
MS-KIF18A, PPP1R99 |
-187.1kb
|
Anti-sense |
Chr. - 28020616 - 28108199 on Build GRCh38 |
Downstream |
microRNA 8068 |
MIR8068 |
hsa-mir-8068 |
181.9kb
|
Anti-sense |
Chr. - 28477481 - 28477548 on Build GRCh38 |
View Full Product Details