Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTO1
Entrez Gene ID
25821
Gene Name
mitochondrial tRNA translation optimization 1
Gene Aliases
CGI-02, COXPD10
Chromosome Location
Chr.6: 73461727 - 73501456 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006715444.2 XP_006715507.1 1 - 2 80
XM_011535724.1 XP_011534026.1 1 - 2 80
XM_017010697.1 XP_016866186.1 1 - 2 80


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA269724.1 - 1 - 2 Chr. 6 - 73461999 - 73473423 [+] on Build GRCh38 80
GenBankmRNA DB034315.1 - 1 - 2 Chr. 6 - 73462084 - 73473532 [+] on Build GRCh38 80
GenBankmRNA DA937826.1 - 1 - 2 Chr. 6 - 73461936 - 73466606 [+] on Build GRCh38 80
GenBankmRNA BP284604.1 - 1 - 2 Chr. 6 - 73461778 - 73466408 [+] on Build GRCh38 80
GenBankmRNA DB042686.1 - 1 - 2 Chr. 6 - 73462084 - 73473520 [+] on Build GRCh38 80
GenBankmRNA DA396214.1 - 1 - 2 Chr. 6 - 73461974 - 73471524 [+] on Build GRCh38 80
GenBankmRNA DQ656037.1 - 2 - 3 296 Chr. 6 - 73462084 - 73466588 [-] on Build GRCh38 80


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream Mab-21 domain containing 1 MB21D1 C6orf150, cGAS, h-cGAS -9.8kb Sense Chr. - 73423711 - 73452330 on Build GRCh38
Downstream eukaryotic translation elongation factor 1 alpha 1 EEF1A1 CCS-3, CCS3, EE1A1, EEF-1, EEF1A, EF-Tu, EF1A, GRAF-1EF, HNGC:16303, LENG7, PTI1, eEF1A-1 49.3kb Sense Chr. - 73515750 - 73521032 on Build GRCh38

View Full Product Details