Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DQ668403.1 - 1 3447 Chr. 15 - 20660950 - 20664970 [+] on Build GRCh38 88


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream golgin A8 family member C, pseudogene GOLGA8CP GOLGA8C -85.2kb Sense Chr. - 20562349 - 20575703 on Build GRCh38
Downstream neurobeachin pseudogene 1 NBEAP1 BCL8, BCL8A 4.5kb Anti-sense Chr. - 20669468 - 20756151 on Build GRCh38

View Full Product Details