Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
EF212256.1 |
- |
1 |
71 |
|
Chr. 11 - 131634146 - 131634281 [+] on Build GRCh38 |
118 |
GenBankmRNA |
AA664089.1 |
- |
1 |
|
|
Chr. 11 - 131633900 - 131634396 [-] on Build GRCh38 |
118 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101929653 |
LOC101929653 |
|
-93.3kb
|
Anti-sense |
Chr. - 131533857 - 131540867 on Build GRCh38 |
Downstream |
NTM antisense RNA 1 |
NTM-AS1 |
C11orf39 |
27.8kb
|
Anti-sense |
Chr. - 131662129 - 131663556 on Build GRCh38 |
View Full Product Details