Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AA077043.1 |
- |
1 |
154 |
|
Chr. 7 - 31703080 - 31703367 [+] on Build GRCh38 |
77 |
GenBankmRNA |
G30739.1 |
- |
1 |
154 |
|
Chr. 7 - 31703080 - 31703367 [+] on Build GRCh38 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
coiled-coil domain containing 129 |
CCDC129 |
|
-44.4kb
|
Sense |
Chr. - 31514071 - 31658720 on Build GRCh38 |
Downstream |
phosphoribosyl pyrophosphate synthetase 2 pseudogene |
LOC100130673 |
|
753.5kb
|
Anti-sense |
Chr. - 32456897 - 32458494 on Build GRCh38 |
View Full Product Details