Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BE644621.1 - 1 401 Chr. 17 - 35824446 - 35824928 [-] on Build GRCh38 102
GenBankmRNA AK022003.1 - 1 1451 Chr. 17 - 35824256 - 35825977 [-] on Build GRCh38 102
GenBankmRNA AW104422.1 - 1 395 Chr. 17 - 35824431 - 35824922 [-] on Build GRCh38 102
GenBankmRNA AW468815.1 - 1 393 Chr. 17 - 35824447 - 35824920 [-] on Build GRCh38 102
GenBankmRNA BF841011.1 - 1 322 Chr. 17 - 35824402 - 35824832 [-] on Build GRCh38 102
GenBankmRNA AA687283.1 - 1 402 Chr. 17 - 35824459 - 35824928 [-] on Build GRCh38 102
GenBankmRNA AW296067.1 - 1 411 Chr. 17 - 35824283 - 35824938 [-] on Build GRCh38 102
GenBankmRNA AA948100.1 - 1 400 Chr. 17 - 35824407 - 35824927 [-] on Build GRCh38 102
GenBankmRNA AW117240.1 - 1 397 Chr. 17 - 35824439 - 35824924 [-] on Build GRCh38 102
GenBankmRNA BF841003.1 - 1 324 Chr. 17 - 35824410 - 35824831 [-] on Build GRCh38 102
GenBankmRNA AI301573.1 - 1 Chr. 17 - 35824256 - 35824688 [+] on Build GRCh38 102
GenBankmRNA AI636891.1 - 1 394 Chr. 17 - 35824386 - 35824921 [-] on Build GRCh38 102
GenBankmRNA BE671762.1 - 1 394 Chr. 17 - 35824386 - 35824921 [-] on Build GRCh38 102
GenBankmRNA BU688925.1 - 1 412 Chr. 17 - 35824225 - 35824939 [-] on Build GRCh38 102
GenBankmRNA AI911145.1 - 1 397 Chr. 17 - 35824448 - 35824924 [-] on Build GRCh38 102
GenBankmRNA AU146490.1 - 1 Chr. 17 - 35824256 - 35824784 [+] on Build GRCh38 102


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream matrix metallopeptidase 28 MMP28 EPILYSIN, MM28, MMP-25, MMP-28, MMP25 -28.5kb Sense Chr. - 35756249 - 35795707 on Build GRCh38
Downstream HEAT repeat containing 9 HEATR9 C17orf66 29.0kb Sense Chr. - 35854946 - 35868891 on Build GRCh38

View Full Product Details