Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SARS
Entrez Gene ID
6301
Gene Name
seryl-tRNA synthetase
Gene Aliases
SERRS, SERS
Chromosome Location
Chr.1: 109213893 - 109238182 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_034072.1 2 - 2 2792 104
GenBank mRNA AK022339.1 - 1 - 1 1850 104


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA N52088.1 - 1 Chr. 1 - 109216900 - 109217379 [-] on Build GRCh38 105
GenBankmRNA BX109203.1 - 1 77 Chr. 1 - 109217026 - 109217487 [+] on Build GRCh38 104
GenBankmRNA AW673148.1 - 1 Chr. 1 - 109216933 - 109217421 [-] on Build GRCh38 104
GenBankmRNA AU147785.1 - 1 Chr. 1 - 109216943 - 109217489 [-] on Build GRCh38 104
GenBankmRNA BF803007.1 - 1 Chr. 1 - 109216855 - 109217272 [-] on Build GRCh38 104
GenBankmRNA DB524918.1 - 1 84 Chr. 1 - 109217019 - 109217488 [+] on Build GRCh38 104
GenBankmRNA AA411730.1 - 1 78 Chr. 1 - 109217026 - 109217230 [+] on Build GRCh38 105
GenBankmRNA AU153741.1 - 1 Chr. 1 - 109216832 - 109217379 [-] on Build GRCh38 104
GenBankmRNA AI991994.1 - 1 Chr. 1 - 109217003 - 109217491 [-] on Build GRCh38 104


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream KIAA1324 KIAA1324 EIG121 -8.5kb Sense Chr. - 109113963 - 109206781 on Build GRCh38
Downstream cadherin EGF LAG seven-pass G-type receptor 2 CELSR2 ADGRC2, CDHF10, EGFL2, Flamingo1, MEGF3 32.2kb Sense Chr. - 109249705 - 109275756 on Build GRCh38

View Full Product Details