Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NKD1
- Entrez Gene ID
-
85407
- Gene Name
- naked cuticle homolog 1
- Gene Aliases
-
Naked1
- Chromosome Location
-
Chr.16: 50548330 - 50649249 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA389991.1 |
- |
1 |
63 |
|
Chr. 16 - 50637783 - 50638379 [+] on Build GRCh38 |
85 |
GenBankmRNA |
AU140394.1 |
- |
1 |
253 |
|
Chr. 16 - 50637593 - 50638043 [+] on Build GRCh38 |
85 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
bromodomain containing 7 |
BRD7 |
BP75, CELTIX1, NAG4 |
-268.6kb
|
Anti-sense |
Chr. - 50317563 - 50368969 on Build GRCh38 |
Downstream |
sorting nexin 20 |
SNX20 |
SLIC1 |
25.6kb
|
Anti-sense |
Chr. - 50666300 - 50681353 on Build GRCh38 |
View Full Product Details