Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LOC101928107
- Entrez Gene ID
-
101928107
- Gene Name
- uncharacterized LOC101928107
- Gene Aliases
-
-
- Chromosome Location
-
Chr.21: 33111704 - 33123700 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_109961.1 |
|
2 - 3 |
327 |
70 |
GenBank mRNA |
BC021745.1 |
- |
3 - 4 |
332 |
70 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG718233.1 |
- |
3 - 4 |
332 |
|
Chr. 21 - 33112321 - 33123700 [-] on Build GRCh38 |
70 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
oligodendrocyte transcription factor 1 |
OLIG1 |
BHLHB6, BHLHE21 |
-39.3kb
|
Anti-sense |
Chr. - 33070144 - 33072422 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 1548 |
LINC01548 |
C21orf54 |
50.9kb
|
Sense |
Chr. - 33165470 - 33170236 on Build GRCh38 |
View Full Product Details