Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB351508.1 - 1 253 Chr. 1 - 77559968 - 77560501 [+] on Build GRCh38 79
GenBankmRNA DB316150.1 - 1 250 Chr. 1 - 77559968 - 77560504 [+] on Build GRCh38 79
GenBankmRNA G36577.1 - 1 248 Chr. 1 - 77559968 - 77560418 [+] on Build GRCh38 79
GenBankmRNA N50729.1 - 1 248 Chr. 1 - 77559968 - 77560418 [+] on Build GRCh38 79


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream adenylate kinase 5 AK5 AK6 Sense Chr. - 77281908 - 77559969 on Build GRCh38
Downstream zinc finger ZZ-type containing 3 ZZZ3 ATAC1 1.1kb Anti-sense Chr. - 77561526 - 77683440 on Build GRCh38

View Full Product Details