Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
R38744.1 |
- |
|
|
|
Chr. - - [-] on Build |
82 |
GenBankmRNA |
AF052176.1 |
- |
1 |
1148 |
|
Chr. 3 - 140056783 - 140058100 [+] on Build GRCh38 |
82 |
GenBankmRNA |
BX092979.1 |
- |
1 |
|
|
Chr. 3 - 140057655 - 140058100 [-] on Build GRCh38 |
82 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
calsyntenin 2 |
CLSTN2 |
ALC-GAMMA, CDHR13, CS2, CSTN2, alcagamma |
|
Sense |
Chr. - 139935185 - 140577397 on Build GRCh38 |
Downstream |
CLSTN2 antisense RNA 1 |
CLSTN2-AS1 |
|
447.5kb
|
Anti-sense |
Chr. - 140505618 - 140508789 on Build GRCh38 |
View Full Product Details