Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI187728.1 - 1 Chr. 17 - 20008055 - 20008263 [+] on Build GRCh38 76
GenBankmRNA HY210207.1 - 1 Chr. 17 - 20008078 - 20008541 [+] on Build GRCh38 76
GenBankmRNA BM670330.1 - 1 Chr. 17 - 20008051 - 20008604 [+] on Build GRCh38 76
GenBankmRNA CN268937.1 - 1 562 Chr. 17 - 20008157 - 20008752 [-] on Build GRCh38 76
GenBankmRNA BC033051.1 - 1 1049 Chr. 17 - 20008051 - 20009234 [-] on Build GRCh38 76


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream A-kinase anchoring protein 10 AKAP10 AKAP-10, D-AKAP-2, D-AKAP2, PRKA10 -30.2kb Sense Chr. - 19904452 - 19977856 on Build GRCh38
Downstream sperm antigen with calponin homology and coiled-coil domains 1 SPECC1 CYTSB, HCMOGT-1, HCMOGT1, NSP 102.0 Anti-sense Chr. - 20009301 - 20319026 on Build GRCh38

View Full Product Details