Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BC035378.1 - 1 755 Chr. 5 - 153901466 - 153903229 [+] on Build GRCh38 86


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream glutamate ionotropic receptor AMPA type subunit 1 GRIA1 GLUH1, GLUR1, GLURA, GluA1, HBGR1 -87.6kb Sense Chr. - 153489615 - 153813873 on Build GRCh38
Downstream family with sequence similarity 114 member A2 FAM114A2 133K02, C5orf3 88.4kb Anti-sense Chr. - 153990131 - 154038938 on Build GRCh38

View Full Product Details