Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA R33702.1 - 124 Chr. - - [-] on Build 79
GenBankmRNA BC067774.1 - 1 1352 Chr. 2 - 237085911 - 237087969 [+] on Build GRCh38 79


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC93463 LOC93463 -29.7kb Anti-sense Chr. - 237048599 - 237056167 on Build GRCh38
Downstream collagen type VI alpha 3 chain COL6A3 BTHLM1, DYT27, UCMD1 236.0kb Anti-sense Chr. - 237324012 - 237434822 on Build GRCh38

View Full Product Details