Assay Design
The assay probe spans an exon junction.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CR608746.1 |
- |
2 - 3 |
144 |
|
Chr. X - 27349150 - 27398989 [-] on Build GRCh38 |
71 |
GenBankmRNA |
BX398782.2 |
- |
2 - 3 |
141 |
|
Chr. X - 27350109 - 27398979 [-] on Build GRCh38 |
71 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105373150 |
LOC105373150 |
|
|
Sense |
Chr. - 27153368 - 27399005 on Build GRCh38 |
Downstream |
protein phosphatase 4 regulatory subunit 3C, pseudogene |
PPP4R3CP |
FLFL3P, SMEK3P, smk1 |
61.4kb
|
Sense |
Chr. - 27460211 - 27463341 on Build GRCh38 |
View Full Product Details