Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCL6
Entrez Gene ID
604
Gene Name
B-cell CLL/lymphoma 6
Gene Aliases
BCL5, BCL6A, LAZ3, ZBTB27, ZNF51
Chromosome Location
Chr.3: 187721377 - 187745725 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005247694.3 XP_005247751.1 1 - 1 2468 75
GenBank mRNA AK090890.1 - 1 - 1 1150 75


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BQ956210.1 - 1 176 Chr. 3 - 187738084 - 187738964 [-] on Build GRCh38 75
GenBankmRNA AV749596.1 - 1 213 Chr. 3 - 187738522 - 187739005 [-] on Build GRCh38 75
GenBankmRNA BG928434.1 - 1 280 Chr. 3 - 187738433 - 187738983 [-] on Build GRCh38 75
GenBankmRNA BQ883719.1 - 1 452 Chr. 3 - 187738341 - 187739240 [-] on Build GRCh38 75
GenBankmRNA AA453841.1 - 1 135 Chr. 3 - 187738498 - 187738923 [-] on Build GRCh38 75
GenBankmRNA AV748863.1 - 1 151 Chr. 3 - 187738570 - 187738939 [-] on Build GRCh38 75
GenBankmRNA BG898348.1 - 1 274 Chr. 3 - 187738657 - 187738983 [-] on Build GRCh38 75


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream hCG1645011-like LOC100131635 -5.4kb Anti-sense Chr. - 187702366 - 187732415 on Build GRCh38
Downstream uncharacterized LOC105374266 LOC105374266 226.9kb Sense Chr. - 187958775 - 187976407 on Build GRCh38

View Full Product Details